Learn More
TNIP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 151041AP
Catalog No. 50-172-9981
Add to cart
Description
Description
Grasp65 is a structural protein of the golgi apparatus. It has been implicated in the stacking of golgi cisternae. Grasp65 is shown to have the properties to bind surfaces together in a mitotically regulated matter.
Specifications
Specifications
TNIP1
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q15025, Q9WUU8
ABIN 1, ABIN1, ABIN-1, HIV 1 Nef interacting protein, hVAN, KIAA0113, NAF1, Nef associated factor 1, Nip40 1, TNFAIP3 interacting protein 1, TNIP1, VAN
Rabbit
IgG
150 μL
-20°C
Primary
10318, 57783
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
TNIP1
Liquid
Tnip1
TNIP1 Fusion Protein Ag1561
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse