Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNIP2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | TNIP2 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 2-4 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056701T
![]() |
Novus Biologicals
NB10056701SS |
0.025 mg |
Each for $181.90
|
|
|||||
NB10056701
![]() |
Novus Biologicals
NB10056701 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
TNIP2 Polyclonal specifically detects TNIP2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
TNIP2 | |
Western Blot 2-4 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS containing 0.05% BSA with 0.05% Sodium Azide | |
A20-binding inhibitor of NF-kappa-B activation 2, A20-binding inhibitor of NF-kappaB activation-2, ABIN-2, ABIN2KLIP, Fetal liver LKB1-interacting protein, FLIP1DKFZp434J1313, MGC4289, TNFAIP3 interacting protein 2, TNFAIP3-interacting protein 2 | |
TNIP2 | |
IgG | |
Protein G purified |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q8NFZ5 | |
79155 | |
This antibody was generated by immunizing rabbits with synthetic peptides of human ABIN-2 (NP_077285). | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title