Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TNIP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528918
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (82%), Rat (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TNFAIP3, also known as A20, is located in chromosome band 6q23, a region that is often deleted in B cell lymphomas. Recently, it was identified as a tumor suppressor gene in Hodgkin lymphoma and several subtypes of non-Hodgkin lymphomas. TNFAIP3 was initially identified as a zinc-finger protein that is rapidly and transiently induced by TNF-a, inhibiting NF-kappa-B-dependent gene expression, and protecting cells from TNF-a-cytotoxicity. Overexpression of TNFAIP3 also inhibits the TLR2- and TLR4-mediated interleukin-8 synthesis in airway epithelial cells, suggesting that TNFAIP3 also acts as a negative regulator of TLR-mediated inflammatory responses, thereby protecting the host against harmful over-responses to pathogens. At least two isoforms of TNFAIP3 are known to exist.
Specifications
TNIP2 | |
Polyclonal | |
Unconjugated | |
Tnip2 | |
1810020H16Rik; A20 binding inhibitor of NF-kappaB activation-2; A20-binding inhibitor of NF-kappa-B activation 2; A20-binding inhibitor of NF-kappaB activation-2; ABIN2; ABIN-2; AI428870; Fetal liver LKB1-interacting protein; FLIP1; KLIP; TNFAIP3 interacting protein 2; TNFAIP3-interacting protein 2; Tnip2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79155 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q8NFZ5 | |
Tnip2 | |
Recombinant fragment corresponding to a region within amino acids 46 and 308 of Human Abin 2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction