Learn More
TNIP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 154591AP
Catalog No. 50-173-0245
Add to cart
Description
Description
TNFAIP3, also known as A20, is located in chromosome band 6q23, a region that is often deleted in B cell lymphomas. Recently, it was identified as a tumor suppressor gene in Hodgkin lymphoma and several subtypes of non-Hodgkin lymphomas. TNFAIP3 was initially identified as a zinc-finger protein that is rapidly and transiently induced by TNF-a, inhibiting NF-kappa-B-dependent gene expression, and protecting cells from TNF-a-cytotoxicity. Overexpression of TNFAIP3 also inhibits the TLR2- and TLR4-mediated interleukin-8 synthesis in airway epithelial cells, suggesting that TNFAIP3 also acts as a negative regulator of TLR-mediated inflammatory responses, thereby protecting the host against harmful over-responses to pathogens. At least two isoforms of TNFAIP3 are known to exist.
Specifications
Specifications
TNIP2
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8NFZ5, Q99JG7
ABIN 2, ABIN2, FLIP1, KLIP, TNFAIP3 interacting protein 2, TNIP2
Rabbit
IgG
150 μL
-20°C
Primary
231130, 305451, 79155
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
TNIP2
Liquid
TNIP2
TNIP2 Fusion Protein Ag7731
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat