Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TNK1 Monoclonal Antibody (170CT7.5.4)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA537485
Description
TNK1 Monoclonal Antibody for Western Blot
TNK1 tyrosine kinase, non-receptor 1 belongs to the tyrosine protein kinase family and as such is an important regulator of intracellular signal transduction pathways mediating cellular proliferation, survival, and development. TNK1 is highly expressed in fetal tissues and at lower levels in a few adult tissues. TNK1 may function in signaling pathways utilized broadly during fetal development, and more selectively in adult tissues. TNK1 plays a negative regulatory role in the Ras-Raf1-MAPK pathway, and knockout mice have been shown to develop spontaneous tumors, suggesting a role as a tumor suppressor gene.
Specifications
TNK1 | |
Monoclonal | |
0.45 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q99ML2 | |
TNK1 | |
TNK1 recombinant protein. | |
400 μL | |
Primary | |
Mouse | |
Antibody | |
IgG1 κ |
Western Blot | |
170CT7.5.4 | |
Unconjugated | |
TNK1 | |
CD38 negative kinase 1; Kinase of embryonic stem cells; Kos1; MGC46193; non-receptor tyrosine kinase; non-receptor tyrosine-protein kinase TNK1; TNK1; Tnk1a; Tnk1b; tyrosine kinase non receptor 1; tyrosine kinase, non-receptor 1; tyrosine kinase, non-receptor, 1 | |
Mouse | |
Protein G | |
RUO | |
83813 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction