Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNK1 Mouse anti-Human, Mouse, Clone: 170CT7.5.4, Abnova™
Mouse monoclonal antibody raised against human TNK1.
Supplier: Abnova Corporation MAB12307
Description
TNK1 is a nonreceptor tyrosine kinase. These kinases, like members of the SRC (MIM 190090) and JAK (see MIM 147795) families, mediate intracellular signaling downstream of receptor activation.[supplied by OMIM]
Specifications
TNK1 | |
Monoclonal | |
Unconjugated | |
Immunohistochemistry (1:50-1:100) Western Blot (1:500) The optimal working dilution should be determined by the end user. | |
MGC46193 | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
170CT7.5.4 | |
Mouse monoclonal antibody raised against human TNK1. | |
In ascites (0.09% sodium azide) | |
TNK1 | |
Recombinant His fusion protein corresponding to human TNK1. | |
RUO | |
8711 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction