Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNN Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550579

Catalog No. PA550579


Add to cart

Description

Description

The antibody detects endogenous levels of total TNN protein.

Involved in neurite outgrowth and cell migration in hippocampal explants.
Specifications

Specifications

TNN
Polyclonal
Unconjugated
TNN
Liquid
TNN
Synthetic peptide corresponding to a region derived from internal residues of human tenascin N
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin)
1.3 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q9UQP3, Q80Z71
TN-W
Rabbit
IgG
100 μL
-20°C
Primary
329278, 63923
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit