Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNNI2 Monoclonal Antibody (7G2), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA122704
Description
MA1-22704 detects skeletal muscle Troponin I in Human samples. MA1-22704 has been successfully used in ELISA and Western Blot procedures. MA1-22704 immunogen corresponds to Full length native protein (purified) (Human)..
This antibody detects Troponin I from human skeletal muscleSpecifications
TNNI2 | |
Monoclonal | |
5.6 mg/mL | |
TNNI2 | |
AMCD2B, DA2B, FSSV, fsTnI | |
Mouse | |
Protein A | |
RUO | |
7136 | |
4° C, do not freeze | |
Liquid |
ELISA, Western Blot | |
7G2 | |
Unconjugated | |
P48788 | |
TNNI2 | |
Full length native protein (purified) (Human). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title