Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TNNI3K Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543126
Description
Peptide sequence: LLKFGADVNV SGEVGDRPLH LASAKGFLNI AKLLMEEGSK ADVNAQDNED Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
TNNI3K may play a role in cardiac physiology.
Specifications
TNNI3K | |
Polyclonal | |
Unconjugated | |
TNNI3K | |
cardiac ankyrin repeat kinase; Cardiac troponin I-interacting kinase; Cark; CCDD; D830019J24Rik; Serine/threonine-protein kinase TNNI3K; si:dkey-267j14.4; TNNI3 interacting kinase; TNNI3-interacting kinase; Tnni3k; tnni3k {ECO:0000250; UniProtKB:Q59H18} | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51086 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q59H18 | |
TNNI3K | |
Synthetic peptide directed towards the N-terminal of human TNNI3K (aa 186-235). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction