Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNNI3K Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | TNNI3K |
---|---|
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB022901
|
Novus Biologicals
NBP294172100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB022902
|
Novus Biologicals
NBP29417220UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
TNNI3K Polyclonal antibody specifically detects TNNI3K in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
TNNI3K | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CARKCardiac ankyrin repeat kinase, EC 2.7.11.1, MGC142099, MGC33828, serine/threonine-protein kinase TNNI3K, TNNI3 interacting kinase, TNNI3-interacting kinase | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-75 of human TNNI3K (NP_057062.1). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Protein Kinase | |
PBS (pH 7.3), 50% glycerol | |
51086 | |
IgG | |
Affinity purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title