Learn More
TNNT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 158931AP
Catalog No. 50-173-0556
Add to cart
Description
Description
The protein encoded by this gene is the tropomyosin-binding subunit of the troponin complex, which is located on the thin filament of striated muscles and regulates muscle contraction in response to alterations in intracellular calcium ion concentration. Mutations in this gene have been associated with familial hypertrophic cardiomyopathy as well as with dilated cardiomyopathy. Transcripts for this gene undergo alternative splicing that results in many tissue-specific isoforms, however, the full-length nature of some of these variants has not yet been determined.
Specifications
Specifications
TNNT1
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O88346, P13805, Q7TNB2
ANM, STNT, TNNT1, TNT, TNTS
Rabbit
IgG
150 μL
-20°C
Primary
171409, 21955, 7138
Western Blot
Unconjugated
TNNT1
Liquid
Tnnt1
TNNT1 Fusion Protein Ag8702
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat