Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNP2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548457

Catalog No. PA548457

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

In the elongating spermatids of mammals, the conversion of nucleosomal chromatin to the compact, non-nucleosomal form found in the sperm nucleus is associated with the appearance of a small set of basic chromosomal transition proteins.
Specifications

Specifications

TNP2
Polyclonal
PBS with 0.09% sodium azide
Q05952
Nuclear transition protein 2, TP-2, TP2, TNP2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7142
Western Blot
Unconjugated
TNP2
Liquid
TNP2
KLH conjugated synthetic peptide between 88-116 amino acids from the C-terminal region of human TNP2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit