Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNRC4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548508

Catalog No. PA548508

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine and mouse based on sequence homology.

RNA-binding protein involved in the regulation of pre- mRNA alternative splicing. Mediates exon inclusion and/or exclusion in pre-mRNA that are subject to tissue-specific and developmentally regulated alternative splicing. Specifically activates exon 5 inclusion of cardiac isoforms of TNNT2 during heart remodeling at the juvenile to adult transition. Activates the splicing of MAPT/Tau exon 10. Binds to muscle-specific splicing enhancer (MSE) intronic sites flanking the alternative exon 5 of TNNT2 pre-mRNA.
Specifications

Specifications

TNRC4
Polyclonal
Unconjugated
CELF3
Liquid
CELF3
KLH conjugated synthetic peptide between 3-32 amino acids from the N-terminal region of human TNRC4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q5SZQ8
CUGBP Elav-like family member 3, CELF-3, Bruno-like protein 1, CAG repeat protein 4, CUG-BP- and ETR-3-like factor 3, ELAV-type RNA-binding protein 1, ETR-1, Expanded repeat domain protein CAG/CTG 4, RNA-binding protein BRUNOL-1, Trinucleotide repeat-containing gene 4 protein, CELF3, BRUNOL1, CAGH4, ERDA4, TNRC4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11189
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit