Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNS3 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP2859690.1ML

Catalog No. NB033898


Add to cart

Description

Description

TNS3 Polyclonal specifically detects TNS3 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

TNS3
Unconjugated
Purified
Rabbit
Immunogen affinity purified
RUO
Primary
64759
Western Blot
PBS, 2% Sucrose
TEM6, TENS1, Tensin 3, tensin-3, Tensin-Like SH2 Domain Containing 1, Tensin-Like SH2 Domain-Containing 1, Tensin-Like SH2 Domain-Containing Protein 1, Thyroid Specific PTB Domain Protein, TPP, Tumor Endothelial Marker 6
The immunogen is a synthetic peptide directed towards the C-terminal region of MOUSE TNS3
0.1ml
Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles.
Polyclonal
Mouse
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit