Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toads Products for Science Education

Bufo marinus.

Specifications

Length (English) 4 to 6 in.
Quantity Each
Length (Metric) 10 to 12.5cm
View More Specs

Products 1
Catalog Number Injection Type Product Type Price Quantity & Availability  
Catalog Number Injection Type Product Type Price Quantity & Availability  
S1713S Products for Science Education
View Documents
Plain Bufo Marinus This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
Description

Description

Dissecting these marine toads can enhance your lesson on amphibians by affording a comparison with preserved frogs. Toads are available plain or injected.

  • Compare and contrast skin, fore and hind feet, and tongue with those of frogs
  • Plain and double injection available; double injection have had latex injected into veins and arteries for easier identification, plain are uncolored
  • Size: 4 to 6 in. (10 to 12.5cm)

Specifications

Specifications

4 to 6 in.
Each
10 to 12.5cm
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit