Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tobramycin sulfate, 50mg/mL in distilled water, sterile-filtered, Alfa Aesar™

Click to view available options
Quantity:
5 x 1mL
79645-27-5
C18H39N5O13S
565.592
MFCD00133864
ZEUUPKVZFKBXPW-UHFFFAOYSA-N
54413
4-amino-2-[4,6-diamino-3-[3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-6-(hydroxymethyl)oxane-3,5-diol;sulfuric acid
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(CC(C(O3)CN)O)N)N.OS(=O)(=O)O
79645-27-5
C18H39N5O13S
565.592
MFCD00133864
ZEUUPKVZFKBXPW-UHFFFAOYSA-N
54413
4-amino-2-[4,6-diamino-3-[3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-6-(hydroxymethyl)oxane-3,5-diol;sulfuric acid
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(CC(C(O3)CN)O)N)N.OS(=O)(=O)O
Physical Form Liquid
Quantity 5 x 1mL
Chemical Name or Material Tobramycin sulfate, 50mg/mL in distilled water, sterile-filtered

RTECSNumber : WK2110000

storageNote1 : -30° to -10°C