Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tobramycin Sulfate, USP, Spectrum™

Click to view available options
Quantity:
5 g
25 g
50 g
500 g
Packaging:
Amber Glass Bottle
Poly Bottle
79645-27-5
C18H39N5O13S
565.59
ZEUUPKVZFKBXPW-UHFFFAOYNA-N
4-amino-2-[(4,6-diamino-3-{[3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy}-2-hydroxycyclohexyl)oxy]-6-(hydroxymethyl)oxane-3,5-diol; sulfuric acid
OS(O)(=O)=O.NCC1OC(OC2C(N)CC(N)C(OC3OC(CO)C(O)C(N)C3O)C2O)C(N)CC1O
79645-27-5
C18H39N5O13S
565.59
ZEUUPKVZFKBXPW-UHFFFAOYNA-N
4-amino-2-[(4,6-diamino-3-{[3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy}-2-hydroxycyclohexyl)oxy]-6-(hydroxymethyl)oxane-3,5-diol; sulfuric acid
OS(O)(=O)=O.NCC1OC(OC2C(N)CC(N)C(OC3OC(CO)C(O)C(N)C3O)C2O)C(N)CC1O

T1598, 79645-27-5, (C18H37N5O9)2•5H2SO4

Spectrum™ Tobramycin Sulfate, USP belongs to a class of drugs known as aminoglycoside antibiotics that works by stopping the growth of bacteria. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Ignition Residue 0.01
Packaging Amber Glass Bottle
Grade USP
Water 0.02
pH 6.0 to 8.0
Quantity 25 g