Tocris™ Toceranib

Potent PDGFR and VEGFR inhibitor

Manufacturer: Tocris™ 3909/50

 View more versions of this product

Catalog No. 39-095-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors Toceranib
    Purity >98
    Molecular Weight 396.46
    Quantity 50mg
    Product Type Toceranib

    Potent ATP-competitive PDGFR and VEGFR inhibitor (Ki = 5 and 6 nM, respectively); inhibits phosphorylation of c-Kit and suppresses the growth of mast cell lines expressing mutant Kit, inducing cell cycle arrest and apoptosis. Also inhibits FGFR1 (Ki = 0.5µM). Effective in vivo.

    • Chemical Name: 5-[(Z)-(5-Fluoro-1,2-dihydro-2-oxo-3H-indol-3-ylidene)methyl]-2,4-dimethyl-N-[2-(1-pyrrolidinyl)ethyl]-1H-pyrrole-3-carboxamide
    • Formula: C22H25FN4O2
    • CAS Number: 356068-94-5
    • Solubility: Soluble to 10mM in DMSO with gentle warming
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit