Learn More
Tocris Bioscience™ 2B-(SP)
Selective GSK-3 phosphopeptide substrate
Manufacturer:  Tocris Bioscience™ 1802/1
Catalog No. 18-021
Add to cart
This item is not returnable. View return policy
Description
Description
Selective phosphopeptide substrate for glycogen synthase kinase-3 (GSK-3); derived from the phosphorylation site of the translation factor eIF2B.