Learn More
Tocris Bioscience™ ADWX 1 Available on GSA/VA Contract for Federal Government customers only.
Potent and selective KV1.3 channel blocker
Manufacturer:  Tocris Bioscience™ 4812/100U
Catalog No. 48-121-00U
Add to cart
This item is not returnable. View return policy
Description
Description
Potent and selective KV1.3 channel blocker (IC50 values are 0.0019 and 0.65 nM for KV1.3 and KV1.1, respectively). Inhibits CD4+ CCR7- T cell activation. Ameliorates rat experimental autoimmune encephalomyelitis, in a model for multiple sclerosis.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
100μg