Learn More
Tocris Bioscience™ AKTide-2T Available on GSA/VA Contract for Federal Government customers only.
Akt/PKB substrate (synthetic)
Manufacturer:  Tocris Bioscience™ 2422/1
Catalog No. 24-221
Add to cart
This item is not returnable. View return policy
Description
Description
Peptide substrate for Akt/PKB. Acts as an inhibitor and mimics the optimal phosphorylation sequence of Akt.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1mg