Learn More
Tocris Bioscience™ [Ala1,3,11,15]-Endothelin
Selective ETB agonist
Manufacturer:  Tocris Bioscience™ 1197/500U
Catalog No. 11975-00U
Add to cart
Description
Description
Selective ET B endothelin receptor agonist (IC 50 values are 0.33 and 2200 nM for displacing [ 125 I]-ET-1 from ET B and ET A receptors respectively).
Specifications
Specifications
500μg