Learn More
Tocris Bioscience™ BIM 187
Bombesin/GRP receptor agonist
Manufacturer:  Tocris Bioscience™ 2716/1
Catalog No. 27-161
Add to cart
This item is not returnable. View return policy
Description
Description
Bombesin/GRP receptor agonist that reduces food intake following i.p. administration.
Specifications
Specifications
1mg