Learn More
Tocris Bioscience™ Catestatin
Non-competitive nicotinic cholinergic antagonist
Supplier:  Tocris Bioscience™ 2722/1
Catalog No. 27-221
Add to Cart
This item is not returnable. View return policy
Description
Description
Non-competitive nAChR antagonist
Catestatin is a non-competitive nicotinic cholinergic antagonist; selectively inhibits nicotinic-stimulated catecholamine secretion from chromaffin cells and noradrenergic neurons (IC50 ∽ 200 nM). Blocks nicotinic-induced cationic signaling (IC50 ∽ 200 - 250 nM) and inhibits nicotinic-agonist induced desensitization of catecholamine release. Also stimulates mast cell release of histamine via a separate mechanism.
Specifications
Specifications
1 mg