Learn More
Tocris Bioscience™ Cyclosomatostatin
Non-selective sst antagonist
Manufacturer:  Tocris Bioscience™ 3493/1
Catalog No. 34-931
Add to cart
This item is not returnable. View return policy
Description
Description
Cyclosomatostatin is a non-selective somatostatin (sst) receptor antagonist. Blocks the effects of sst on airway β-adrenergic function, CRF-induced suppression of gastric empyting, modulation of ACh release and growth hormone, insulin and glucagon release. Reported to act as an sst receptor agonist in human neuroblastoma cell line SH-SY5Y.
Specifications
Specifications
1 mg