Learn More
Tocris Bioscience™ Deltorphin I
Potent δ agonist
Manufacturer:  Tocris Bioscience™ 4591/1
Catalog No. 45-911
Add to cart
This item is not returnable. View return policy
Description
Description
Potent δ-opioid receptor agonist (IC50 = 0.5 nM in human granulocytes). Exhibits a high rate of blood-brain barrier penetrance.
Specifications
Specifications
1mg