Learn More
Tocris Bioscience™ Endomorphin-2
Potent and selective μ agonist
Manufacturer:  Tocris Bioscience™ 1056/5
Catalog No. 10-565
Add to cart
This item is not returnable. View return policy
Description
Description
Endomorphin-2 is a endogenous peptide with extremely high affinity and selectivity for μ-opioid receptors (with Ki values of 0.69, 9233 and 5240 nM for μ, δ and κ respectively).
Specifications
Specifications
5mg