Learn More
Tocris Bioscience™ Eptifibatide Available on GSA/VA Contract for Federal Government customers only.
Glycoprotein IIb/IIIa inhibitor; inhibits platelet aggregation
Manufacturer:  Tocris Bioscience™ 4725/1
Catalog No. 47-251
Add to cart
This item is not returnable. View return policy
Description
Description
Glycoprotein (GP) IIb/IIIa inhibitor. Reversibly and competitively binds to GPIIa/IIIb on platelets, inhibiting aggregation.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1mg