Learn More
Tocris Bioscience™ Galanin (1-30) (human)
Endogenous galanin receptor agonist
Manufacturer:  Tocris Bioscience™ 1179/200U
Catalog No. 11792-00U
Add to cart
Description
Description
Endogenous peptide with multiple endocrine, metabolic and behavioral effects. Has been shown to have an action on intestinal smooth muscle, insulin and somatostatin release, and synaptic neurotransmission.
Specifications
Specifications
200μg