Learn More
Tocris Bioscience™ GaTx2 Available on GSA/VA Contract for Federal Government customers only.
High affinity, selective ClC-2 blocker; inhibits slow-gating
Manufacturer:  Tocris Bioscience™ 4911/100U
Catalog No. 49-111-00U
Add to cart
This item is not returnable. View return policy
Description
Description
Very high affinity ClC-2 blocker (apparent KD ~ 50 pM). Slows ClC-2 activation and inhibits slow-gating but does not inhibit open ClC-2 channels. Selective for ClC-2 over other ClC family members (ClC-0, ClC-1, ClC-3 and ClC-4)TR, GABAC, CaCC and KV1.2.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
100μg