Learn More
Tocris Bioscience™ GLP-2 (rat) Available on GSA/VA Contract for Federal Government customers only.
Endogenous hormone; displays intestinotrophic activity
Manufacturer:  Tocris Bioscience™ 2259/1
Catalog No. 22-591
Add to cart
This item is not returnable. View return policy
Description
Description
Endogenous peptide identified as an intestinal epithelium-specific growth factor; stimulates cell proliferation and inhibits apoptosis. Diverse effects on gastrointestinal function including regulation of intestinal glucose transport, food intake, and gastric acid secretion.
GLP-2 (human) also available.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1mg