Learn More
Tocris Bioscience™ α-helical CRF 9-41
CRF receptor antagonist
Manufacturer:  Tocris Bioscience™ 1184/500U
Catalog No. 11845-00U
Add to cart
Description
Description
Corticotropin-releasing factor receptor antagonist (K i values are 17, 5 and 0.97 at human CRF 1 rat CRF 2α and mouse CRF 2β receptors respectively).
Specifications
Specifications
500μg