Learn More
Tocris Bioscience™ Huwentoxin IV Available on GSA/VA Contract for Federal Government customers only.
Selective NaV1.7 channel blocker
Manufacturer:  Tocris Bioscience™ 4718/100U
Catalog No. 47-181-00U
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
Selective NaV1.7 channel blocker. Preferentially inhibits neuronal NaV1.7, 1.2 and 1.3 (IC50 values are 26, 150 and 338 nM respectively), compared to muscle subtypes NaV1.4 and 1.5 (IC50 = >10 μM). Inhibits the channel by binding at the neurotoxin receptor site 4 in the S3-S4 linker of domain II, trapping the voltage sensor in the inward, closed configuration.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
100μg