Learn More
Tocris Bioscience™ IRL-1620 Available on GSA/VA Contract for Federal Government customers only.
Highly selective ETB agonist
Manufacturer:  Tocris Bioscience™ 1196/500U
Catalog No. 11965-00U
Add to cart
Description
Description
Highly selective ET B endothelin receptor agonist (K i values are 0.016 and 1900 nM at ET B and ET A receptors respectively). Discriminates two subpopulations of ET B receptors.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
500μg