Learn More
Tocris Bioscience™ JKC 363
Potent and selective MC4 receptor antagonist
Manufacturer:  Tocris Bioscience™ 3426/500U
Catalog No. 34-265-00U
Add to cart
This item is not returnable. View return policy
Description
Description
JKC 363 is a potent and selective melanocortin MC4 receptor antagonist (IC50 values are 0.5 and 44.9 nM at MC4 and MC3 respectively) that antagonizes the effects of α-MSH. Suppresses thyrotropin-releasing hormone (TRH) release, attenuates food intake and reduces formalin-induced pain in vivo.
Specifications
Specifications
500 μg