Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LDV FITC

Fluorescent ligand for α4β1 (VLA-4)

Manufacturer:  Tocris Bioscience™ 4577/1

Catalog No. 45-771


Add to cart
This item is not returnable. View return policy

Description

Description

LDV FITC is a fluorescent ligand that binds to the α4β1 integrin (VLA-4) with high affinity (Kd values are 0.3 nM and 12 nM for binding to U937 cells in the presence and absence of Mn2+ respectively). Used to detect VLA-4 affinity and conformational changes.

Specifications

Specifications

M.W. (Observed): 1368.54
1 mg
Store at -20°C
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit