Learn More
Tocris Bioscience™ Leuprolide Available on GSA/VA Contract for Federal Government customers only.
GnRH receptor agonist
Manufacturer:  Tocris Bioscience™ 2873/1
Catalog No. 28-731
Add to cart
This item is not returnable. View return policy
Description
Description
Gonadotropin-releasing hormone (GnRH, luteinizing hormone-releasing hormone, LHRH) receptor agonist. Suppresses estradiol, LH and FSH serum levels and represses the growth of experimental rat endometriosis.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1mg