Learn More
Tocris Bioscience™ Nafarelin Available on GSA/VA Contract for Federal Government customers only.
GnRH receptor agonist
Manufacturer:  Tocris Bioscience™ 2544/1
Catalog No. 25-441
Add to cart
This item is not returnable. View return policy
Description
Description
Synthetic gonadotropin-releasing hormone (GnRH) agonist that stimulates secretion of luteinizing hormone (LH) and follicle-stimulating hormone (FSH). Repeated administration causes desensitization of pituitary GnRH receptors resulting in inhibition of gonadotropin release and decreased steroid hormone synthesis. Induces atrophy and regression of experimentally induced endometriosis in rat and shrinks hypertrophic tissue in benign prostatic hyperplasia.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1mg