Learn More
Tocris Bioscience™ Neurokinin A (porcine)
Endogenous tachykinin peptide
Manufacturer:  Tocris Bioscience™ 1152/1
Catalog No. 11-521
Add to cart
Description
Description
Endogenous tachykinin peptide; more potent bronchoconstrictor than Substance P.
Specifications
Specifications
1mg