Learn More
Tocris Bioscience™ Neuromedin U (rat) Available on GSA/VA Contract for Federal Government customers only.
Endogenous modulator of blood pressure and flow, gut ion transport, feeding and body temperature
Manufacturer:  Tocris Bioscience™ 1917/1
Catalog No. 19-171
Add to cart
This item is not returnable. View return policy
Description
Description
Endogenous peptide regulating blood pressure, ion transport in the gut, mesenteric blood flow and adrenocortical function. Also decreases food intake and body weight, increases body temperature and heat production, and inhibits gastric acid secretion via the CRH system following central administration in vivo.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1mg