Learn More
Tocris Bioscience™ NocII Available on GSA/VA Contract for Federal Government customers only.
Orphan neuropeptide
Manufacturer:  Tocris Bioscience™ 1119/1
Catalog No. 11-191
Add to cart
This item is not returnable. View return policy
Description
Description
Orphan neuropeptide, flanking nociceptin in the precursor pronociceptin. Stimulates locomotion in mice, possibly through a novel specific receptor.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1mg