Learn More
Tocris Bioscience™ Nocistatin (bovine) Available on GSA/VA Contract for Federal Government customers only.
Opposes action of nociceptin
Manufacturer:  Tocris Bioscience™ 1118/1
Catalog No. 11-181
Add to cart
This item is not returnable. View return policy
Description
Description
An endogenous peptide derived from the same precursor as nociceptin, which is able to block nociceptin-induced allodynia and hyperalgesia and may play an opposing role in pain transmission. Shown to reverse nociceptin inhibition of glutamate release from rat brain slices; also modulates 5-HT transmission in the mouse neocortex via mechanisms separate from nociceptin/orphanin FQ.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1mg