Learn More
Tocris Bioscience™ P11 Available on GSA/VA Contract for Federal Government customers only.
Potent antagonist of αvβ3-vitronectin interaction; antiangiogenic
Manufacturer:  Tocris Bioscience™ 4744/1
Catalog No. 47-441
Add to cart
This item is not returnable. View return policy
Description
Description
Potent antagonist of the integrin αvβ3-vitronectin interaction (IC50 = 25.72 nM). Blocks proliferation and induces apoptosis in HUVECs; antiangiogenic.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1mg