Learn More
Tocris Bioscience™ PBP 10 Available on GSA/VA Contract for Federal Government customers only.
Selective FPR2 antagonist; cell permeable
Manufacturer:  Tocris Bioscience™ 4611/1
Catalog No. 46-111
Add to cart
This item is not returnable. View return policy
Description
Description
Selective formyl peptide receptor 2 (FPR2) antagonist; cell permeable. Selectively inhibits FPR2-mediated NADPH oxidase activity but has no effect on FPR1 signaling in neutrophils. Displays PIP2 binding activity in vitro and blocks cell motility. Also exhibits antiviral activity against influenza viruses via inhibition of viral-induced ERK activation.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1mg