Learn More
Tocris Bioscience™ PG 106
Selective MC3 receptor antagonist
Manufacturer:  Tocris Bioscience™ 4919/1
Catalog No. 49-191
Add to cart
This item is not returnable. View return policy
Description
Description
Selective hMC3 receptor antagonist (IC50 = 210 nM). Exhibits no activity at hMC4 receptors and hMC5 receptors.
Specifications
Specifications
1mg