Learn More
Tocris Bioscience™ [Phe1ψ(CH2-NH)Gly2]Nociceptin(1-13)NH2
Selective nociceptin receptor agonist
Manufacturer:  Tocris Bioscience™ 1092/1
Catalog No. 10-921
Add to cart
This item is not returnable. View return policy
Description
Description
[Phe1Ψ(CH2-NH)Gly2]Nociceptin(1-13)NH2 is a potent agonist of the nociceptin (ORL1) receptor, demonstrated both in vitro and in vivo. Selective, competitive antagonism at the nociceptin receptor has also been reported (pA2 = 7.02 and 6.75 in the guinea pig ileum and mouse vas deferens respectively).
Specifications
Specifications
1mg