Learn More
Tocris Bioscience™ ProTx I Available on GSA/VA Contract for Federal Government customers only.
CaV3.1 channel blocker; also inhibits NaV1.8 and KV2.1
Manufacturer:  Tocris Bioscience™ 4665/100U
Catalog No. 46-651-00U
Add to cart
This item is not returnable. View return policy
Description
Description
Selective CaV3.1 channel blocker (IC50 values are 0.2 and 31.8 μM for hCaV3.1 and hCaV3.2 respectively). Also reversibly inhibits NaV1.8 and blocks KV2.1 channels.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
100μg