Learn More
Tocris Bioscience™ R8-T198wt Available on GSA/VA Contract for Federal Government customers only.
Pim-1 kinase inhibitor
Manufacturer:  Tocris Bioscience™ 4592/1
Catalog No. 45-921
Add to cart
This item is not returnable. View return policy
Description
Description
Cell-permeable peptide inhibitor of Pim-1 kinase, derived from p27Kip1. Inhibits Pim-1 phosphorylation of p27Kip1 and Bad; induces cell cycle arrest (at G1) and apoptosis in DU145 prostate cancer cells. Also inhibits Pim-1-dependent growth of DU145 cells in vitro and in vivo. Displays no effect on the growth of normal prostate epithelial RPWE-1 cells at concentrations of 10 and 20 μM.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1mg