Learn More
Tocris Bioscience™ Rac1 Inhibitor F56, control peptide Available on GSA/VA Contract for Federal Government customers only.
Control peptide version of Rac1 Inhibitor W56 (Cat. No. 2221)
Manufacturer:  Tocris Bioscience™ 2222/1
Catalog No. 22-221
Add to cart
This item is not returnable. View return policy
Description
Description
Control peptide version of Rac1 Inhibitor; comprises residues 45-60 of Rac1 with Trp56 replaced by Phe. Does not affect GEF-Rac1 interaction.
Active Analog also available.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1mg
Rho GTPase Inhibitors