Learn More
Tocris Bioscience™ Spinorphin Available on GSA/VA Contract for Federal Government customers only.
Enkephalin-degrading and ACE inhibitor
Manufacturer:  Tocris Bioscience™ 2931/1
Catalog No. 29-311
Add to cart
This item is not returnable. View return policy
Description
Description
Endogenous peptide; inhibits enkephalin-degrading enzymes (aminopeptidase, dipeptidyl aminopeptidase III, neprilysin) and angiotensin-converting enzyme. Displays antinociceptive effects in mice.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1mg